Pimelic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Heptanedioic acid, 1,5-Pentanedicarboxylic acid, 1,7-Heptanedioic acid
M-Clarity™ quality level = MQ200
P45001-100G P45001-25G
SIALP45001-100GEA 185 EUR
SIALP45001-100G SIALP45001-25G
Pimelic acid, Sigma-Aldrich®
Pimelic acid
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 101…103 °C
Density: 1.329 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004425
CAS Number: 111-16-0
EINECS: 203-840-8
Merck Index: 12,07585