Suberic acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1,8-Octanedioic acid, Octanedioic acid, 1,6-Hexanedicarboxylic acid

M-Clarity™ quality level = MQ200

60930-100G 60930-1KG 60930-250G
SIAL60930-100GEA 65.1 EUR
SIAL60930-100G SIAL60930-1KG SIAL60930-250G
Suberic acid, Sigma-Aldrich®
Suberic acid
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 139…142 °C
Density: 1.354 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004428
CAS Number: 505-48-6
EINECS: 208-010-9
Merck Index: 12,09031

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR