Azelaic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid
M-Clarity™ quality level = MQ200
11480-1KG 11480-250G
SIAL11480-1KGEA 160 EUR
SIAL11480-1KG SIAL11480-250G
Azelaic acid, Sigma-Aldrich®
Azelaic acid
Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 106 °C
Density: 1.0287 g/cm³ (20 °C)
Flash Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00004432
CAS Number: 123-99-9
EINECS: 204-669-1
Merck Index: 13,00908