Azelaic acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid

M-Clarity™ quality level = MQ200

246379-100G 246379-25G
SIAL246379-100GEA 151 EUR
SIAL246379-100G SIAL246379-25G
Azelaic acid, Sigma-Aldrich®
Azelaic acid
Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 106 °C
Density: 1.0287 g/cm³ (20 °C)
Flash Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00004432
CAS Number: 123-99-9
EINECS: 204-669-1
Merck Index: 13,00908

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR