di-Potassium oxalate monohydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Oxalic acid potassium salt monohydrate, Potassium oxalate hydrate, Potassium oxalate monohydrate, Potassium ethanedioate monohydrate
M-Clarity™ quality level = MQ200
P0963-100G P0963-500G
SIALP0963-100GEA 47.1 EUR
SIALP0963-100G SIALP0963-500G
di-Potassium oxalate monohydrate, Sigma-Aldrich®
di-Potassium oxalate monohydrate
Formula: (COOK)₂·H₂O
Melting Pt: 356 °C
Density: 2.154 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150033
CAS Number: 6487-48-5
EINECS: 209-506-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,07820