Disodium oxalate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances
M-Clarity™ quality level = MQ100
379735-25G 379735-5G
SIAL379735-25GEA 220 EUR
SIAL379735-25G SIAL379735-5G
Disodium oxalate, Sigma-Aldrich®
di-Sodium oxalate
Formula: Na₂C₂O₄
MW: 134 g/mol
Melting Pt: 250…270 °C
Density: 2.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
Merck Index: 13,08723