Potassium citrate monohydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate
M-Clarity™ quality level = MQ200
25107-1KG 25107-25KG
SIAL25107-1KGEA 74.1 EUR
SIAL25107-1KG SIAL25107-25KG
Potassium citrate monohydrate, Sigma-Aldrich®
Potassium citrate monohydrate
Formula: HOC(COOK)(CH₂COOK)₂·H₂O
MW: 324.41 g/mol
Melting Pt: 230
Density: 1.98 g/cm³ (20 °C)
MDL Number: MFCD00150442
CAS Number: 6100-05-6
EINECS: 212-755-5
Merck Index: 13,07706
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc3e6bae942870a760ea6cb418203db7458ff3dc&vid=2c2580a7bfc83e60c4dfca9f8af4ac3b3db56b27&prid=web&clid=SAPDAM