Potassium citrate monohydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
C8385-1KG C8385-500G
SIALC8385-1KGEA 83.2 EUR
SIALC8385-1KG SIALC8385-500G
Potassium citrate monohydrate, Sigma-Aldrich®
Potassium citrate monohydrate
Formula: HOC(COOK)(CH₂COOK)₂·H₂O
Melting Pt: 230 °C
Density: 1.98 g/cm³ (20 °C)
MDL Number: MFCD00150442
CAS Number: 6100-05-6
EINECS: 212-755-5
Merck Index: 13,07706
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc3e6bae942870a760ea6cb418203db7458ff3dc&vid=2c2580a7bfc83e60c4dfca9f8af4ac3b3db56b27&prid=web&clid=SAPDAM