Diammonium hydrogen citrate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL25102-6X1KG-REA 431 EUR
SIAL25102-6X1KG-R
Diammonium hydrogen citrate, Sigma-Aldrich®
Diammonium hydrogen citrate
Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226,19 g/mol
Density: 1,483 g/cm³ (20 °C)
MDL Number: MFCD00013068
CAS Number: 3012-65-5
Merck Index: 12,00540

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR