Diammonium hydrogen citrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIAL247561-2.5KGEA
164
EUR
SIAL247561-2.5KG
SIAL247561-100G
SIAL247561-500G
Diammonium hydrogen citrate, Sigma-Aldrich®
Diammonium hydrogen citrate
Formula:
HOC(CO₂H)(CH₂CO₂NH₄)₂ MW: 226,19 g/mol Density: 1,483 g/cm³ (20 °C) |
MDL Number:
MFCD00013068 CAS Number: 3012-65-5 Merck Index: 12,00540 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...