Thioacetic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
T30805-100G T30805-500G T30805-5G
SIALT30805-100GEA 62.7 EUR
SIALT30805-100G SIALT30805-500G SIALT30805-5G
Thioacetic acid, Sigma-Aldrich®
Thioacetic acid
Formula: C₂H₄OS
MW: 76.12 g/mol
Boiling Pt: 90
Melting Pt: –17 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 18
MDL Number: MFCD00004853
CAS Number: 507-09-5
EINECS: 208-063-8
UN: 2436
ADR: 3 II
Merck Index: 12,09454