Thioacetamide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Ethanethioamide
172502-100G 172502-25G 172502-500G
SIAL172502-100GEA 76 EUR
SIAL172502-100G SIAL172502-25G SIAL172502-500G
Thioacetamide, Sigma-Aldrich®
Thioacetamide
Formula: CH₃CSNH₂
MW: 75.13 g/mol
Melting Pt: 113…114 °C
Density: 1.336 g/cm³ (20 °C)
MDL Number: MFCD00008070
CAS Number: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM