Perfluorooctane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Octadecafluorooctane
M-Clarity™ quality level = MQ200
359238-100G 359238-25G
SIAL359238-100GEA 221 EUR
SIAL359238-100G SIAL359238-25G
Perfluorooctane, Sigma-Aldrich®
Perfluorooctane
Formula: C₈F₁₈
MW: 438.06 g/mol
Boiling Pt: 102
Melting Pt: –25 °C
Density: 1.73 g/cm³ (20 °C)
MDL Number: MFCD00042083
CAS Number: 307-34-6
EINECS: 206-199-2