Trifluoroacetamide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2,2,2-Trifluoroacetamide
M-Clarity™ quality level = MQ100
144657-100G 144657-25G
SIAL144657-100GEA 105 EUR
SIAL144657-100G SIAL144657-25G
Trifluoroacetamide, Sigma-Aldrich®
Trifluoroacetamide
Formula: C₂H₂F₃NO
MW: 113.04 g/mol
Boiling Pt: 162.4 °C (1013 hPa)
Melting Pt: 73…75 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00008008
CAS Number: 354-38-1
EINECS: 206-559-9