Methyl difluoroacetate, Sigma-Aldrich®
Supplier: Merck
MQ100
295914-25G
295914-5G
SIAL295914-25GEA
86.9
EUR
SIAL295914-25G
SIAL295914-5G
Methyl difluoroacetate, Sigma-Aldrich®
Methyl difluoroacetate
Formula:
C₃H₄F₂O₂ MW: 110.06 g/mol Boiling Pt: 84…85 °C (1013 hPa) Density: 1.272 g/cm³ (20 °C) |
MDL Number:
MFCD00039276 CAS Number: 433-53-4 EINECS: 207-089-7 UN: 2924 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...