Methyl trifluoroacetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
249831-100G 249831-25G
SIAL249831-100GEA 156 EUR
SIAL249831-100G SIAL249831-25G
Methyl trifluoroacetate, Sigma-Aldrich®
Methyl trifluoroacetate
Formula: C₃H₃F₃O₂
Boiling Pt: 43 °C (1013 hPa)
Melting Pt: –78 °C
Density: 1.2639 g/cm³ (20 °C)
MDL Number: MFCD00000417
CAS Number: 431-47-0
EINECS: 207-074-5
UN: 2924
ADR: 3,II