Difluoroacetic acid, Sigma-Aldrich®

Supplier: Merck
Danger


MQ200

142859-25G 142859-5G
SIAL142859-25GEA 206 EUR
SIAL142859-25G SIAL142859-5G
Difluoroacetic acid, Sigma-Aldrich®
Difluoroacetic acid
Formula: C₂H₂F₂O₂
MW: 96.03 g/mol
Boiling Pt: 134
Melting Pt: –1 °C
Density: 1.526 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004220
CAS Number: 381-73-7
EINECS: 206-839-0
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR