1,1,1-Trifluoroiodoethane 99%
Supplier: Thermo Scientific
Warning
Synonyms: 1-Iodo-2,2,2-trifluoroethane, 2,2,2-Trifluoroethyliodide, 2-Iodo-1,1,1-trifluoroethane, 2,2,2-Trifluoroiodoethane, 1,1,1-Trifluoro-2-iodoethane
176080100 176080500
ACRO176080100EA 60.7 EUR
ACRO176080100 ACRO176080500
1,1,1-Trifluoroiodoethane 99%
1,1,1-Trifluoroiodoethane
Formula: C₂H₂F₃I
MW: 209.94 g/mol
Boiling Pt: 53…55 °C (760 mmHg)
Density: 2.13 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00001082
CAS Number: 353-83-3
EINECS: 206-541-0
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless to pink Liquid
Infrared spectrum Conforms
Refractive index 1.4000 to 1.4020 (20°C, 589 nm)
GC ≥98.5 %
Iron (Fe) ≤0.02 %
Heavy metals (as Pb) ≤0.003 %