1,1,1-Trifluoroiodoethane, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Iodo-2,2,2-trifluoroethane, 2,2,2-Trifluoroethyliodide, 2-Iodo-1,1,1-trifluoroethane, 2,2,2-Trifluoroiodoethane, 1,1,1-Trifluoro-2-iodoethane
M-Clarity™ quality level = MQ100
177814-25G 177814-5G
SIAL177814-25GEA 161 EUR
SIAL177814-25G SIAL177814-5G
1,1,1-Trifluoroiodoethane, Sigma-Aldrich®
1,1,1-Trifluoroiodoethane
Formula: C₂H₂F₃I
MW: 209.94 g/mol
Boiling Pt: 54.7 °C (1013 hPa)
Density: 2.1499 g/cm³ (20 °C)
MDL Number: MFCD00001082
CAS Number: 353-83-3
EINECS: 206-541-0
UN: 2810
ADR: 6.1,III