1-Amino-2,2,2-trifluoroethane, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2,2,2-Trifluoroethylamine
M-Clarity™ quality level = MQ100
269042-1G
269042-5G
SIAL269042-1GEA
57.9
EUR
SIAL269042-1G
SIAL269042-5G
1-Amino-2,2,2-trifluoroethane, Sigma-Aldrich®
1-Amino-2,2,2-trifluoroethane
Formula:
C₂H₄F₃N MW: 99.06 g/mol Boiling Pt: 35.85 Density: 1.2452 g/cm³ (25 °C) |
MDL Number:
MFCD00008132 CAS Number: 753-90-2 EINECS: 212-041-3 UN: 2733 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...