Trifluoroacetic anhydride

Supplier: Roth Carl

ROTH0027.1EA 116 EUR
ROTH0027.1
Trifluoroacetic anhydride
Trifluoroacetic anhydride
Formula: C₄F₆O₃
MW: 210,03 g/mol
Boiling Pt: 39,4 °C (1013 hPa)
Melting Pt: –65 °C
Density: 1,487 g/cm³ (20 °C)
MDL Number: MFCD00000416
UN: 3265
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR