1,1,1,3,3,3-Hexafluoro-2-propanol, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP
M-Clarity™ quality level = MQ200
105228-100G
105228-25G
105228-500G
105228-5G
SIAL105228-100GEA
278
EUR
SIAL105228-100G
SIAL105228-25G
SIAL105228-500G
SIAL105228-5G
1,1,1,3,3,3-Hexafluoro-2-propanol, Sigma-Aldrich®
1,1,1,3,3,3-Hexafluoro-2-propanol
Formula:
C₃H₂F₆O MW: 168.04 g/mol Boiling Pt: 59.1 Melting Pt: –96 °C Density: 1.492 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011651 CAS Number: 920-66-1 EINECS: 213-059-4 UN: 1760 ADR: 8 II Merck Index: 13,09598 |