Triisopropylsilyl trifluoromethanesulphonate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Trifluoromethanesulphonic acid triisopropylsilyl ester, Tri(propan-2-yl)silyl trifluoromethanesulfonate
M-Clarity™ quality level = MQ100
248460-10G
248460-50G
SIAL248460-10GEA
127
EUR
SIAL248460-10G
SIAL248460-50G
Triisopropylsilyl trifluoromethanesulphonate, Sigma-Aldrich®
Triisopropylsilyl trifluoromethanesulphonate
Formula:
C₁₀H₂₁F₃O₃SSi Boiling Pt: 120 °C (1013 hPa) Density: 1.136 g/cm³ (20 °C) |
MDL Number:
MFCD00009913 CAS Number: 80522-42-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...