Methyl trifluoromethanesulphonate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
164283-10G 164283-50G
SIAL164283-10GEA 75.9 EUR
SIAL164283-10G SIAL164283-50G
Methyl trifluoromethanesulphonate, Sigma-Aldrich®
Methyl trifluoromethanesulphonate
Formula: C₂H₃F₃O₃S
MW: 164.11 g/mol
Boiling Pt: 95 °C (1013 hPa)
Density: 1.45 g/cm³ (20 °C)
MDL Number: MFCD00000409
CAS Number: 333-27-7
EINECS: 206-371-7