Trifluoromethanesulphonic anhydride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Triflic anhydride
M-Clarity™ quality level = MQ200
91737-100ML
91737-25ML
91737-5ML
SIAL91737-100MLEA
1030
EUR
SIAL91737-100ML
SIAL91737-25ML
SIAL91737-5ML
Trifluoromethanesulphonic anhydride, Sigma-Aldrich®
Trifluoromethanesulphonic anhydride
Formula:
C₂F₆O₅S₂ MW: 282.14 g/mol Boiling Pt: 83 °C (1013 hPa) Density: 1.67 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000408 CAS Number: 358-23-6 EINECS: 206-616-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...