1,2-Diiodoethane, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
D122807-100G D122807-25G
SIALD122807-100GEA 185 EUR
SIALD122807-100G SIALD122807-25G
1,2-Diiodoethane, Sigma-Aldrich®
1,2-Diiodoethane
Formula: C₂H₄I₂
MW: 281.86 g/mol
Boiling Pt: 206
Melting Pt: 80…82 °C
Density: 2.132 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001093
CAS Number: 624-73-7
EINECS: 210-859-5