1-Iodobutane, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1-Butyl iodide
M-Clarity™ quality level = MQ100
167304-100G
167304-500G
167304-5G
SIAL167304-100GEA
39.9
EUR
SIAL167304-100G
SIAL167304-500G
SIAL167304-5G
1-Iodobutane, Sigma-Aldrich®
1-Iodobutane
Formula:
C₄H₉I MW: 184.02 g/mol Boiling Pt: 130 °C (1013 hPa) Melting Pt: –103 °C Density: 1.616 g/cm³ (20 °C) Flash Pt: 36 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001098 CAS Number: 542-69-8 EINECS: 208-824-4 UN: 1993 ADR: 3,III Merck Index: 13,01572 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...