2-Iodopropane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Isopropyl iodide
M-Clarity™ quality level = MQ200
148938-100G 148938-500G 148938-5G
SIAL148938-100GEA 50.9 EUR
SIAL148938-100G SIAL148938-500G SIAL148938-5G
2-Iodopropane, Sigma-Aldrich®
2-Iodopropane
Formula: C₃H₇I
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: 2.8 °C
Density: 1.7 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00001071
CAS Number: 75-30-9
EINECS: 200-859-3
UN: 2392
ADR: 3,III
Merck Index: 13,05234