Iodoethane, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Ethyl iodide

M-Clarity™ quality level = MQ200

I7780-100G I7780-500G I7780-5G
SIALI7780-100GEA 45.4 EUR
SIALI7780-100G SIALI7780-500G SIALI7780-5G
Iodoethane, Sigma-Aldrich®
Iodoethane
Formula: C₂H₅I
MW: 155.97 g/mol
Boiling Pt: 72 °C (1013 hPa)
Melting Pt: –108 °C
Density: 1.936 g/cm³ (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00001091
CAS Number: 75-03-6
EINECS: 200-833-1
UN: 2810
ADR: 6.1,III
Merck Index: 13,03846

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR