Iodomethane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Methyl iodide
M-Clarity™ quality level = MQ200
I8507-100ML I8507-1L I8507-500ML I8507-5ML
SIALI8507-100MLEA 132 EUR
SIALI8507-100ML SIALI8507-1L SIALI8507-500ML SIALI8507-5ML
Iodomethane, Sigma-Aldrich®
Iodomethane
Formula: CH₃I
MW: 141.94 g/mol
Boiling Pt: 42.4
Melting Pt: –66 °C
Density: 2.279 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001073
CAS Number: 74-88-4
EINECS: 200-819-5
UN: 2644
ADR: 6.1 I
Merck Index: 12,06161