2-Iodoethanol, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

176850-25G 176850-5G
SIAL176850-25GEA 216 EUR
SIAL176850-25G SIAL176850-5G
2-Iodoethanol, Sigma-Aldrich®
2-Iodoethanol
Formula: C₂H₅IO
Boiling Pt: 85 °C (1013 hPa)
Density: 2.202 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002830
CAS Number: 624-76-0
EINECS: 210-861-6
UN: 2810
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR