Triethanolamine (trolamine), Sigma-Aldrich®
Supplier: Merck
Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol
90279-100ML 90279-500ML 90279-2.5L
SIAL90279-100MLEA 21.1 EUR
SIAL90279-100ML SIAL90279-500ML SIAL90279-2.5L
Triethanolamine (trolamine), Sigma-Aldrich®
Triethanolamine (Trolamine)
Formula: C₆H₁₅NO₃
Boiling Pt: 335.4 °C (1013 hPa)
Melting Pt: 21.6 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 179 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM