Triethylamine, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
T0886-100ML T0886-1L T0886-500ML
SIALT0886-100MLEA 24 EUR
SIALT0886-100ML SIALT0886-1L SIALT0886-500ML
Triethylamine, Sigma-Aldrich®
Triethylamine
Formula: (C₂H₅)₃N
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.729 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3,II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM