N,N-Dimethyloctylamine, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: N,N-Dimethyloctan-1-amine

M-Clarity™ quality level = MQ200

256226-25ML 256226-5ML
SIAL256226-25MLEA 92.6 EUR
SIAL256226-25ML SIAL256226-5ML
N,N-Dimethyloctylamine, Sigma-Aldrich®
N,N-Dimethyloctylamine
Formula: C₁₀H₂₃N
MW: 157.3 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: –57 °C
Density: 0.765 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00009558
CAS Number: 7378-99-6
EINECS: 230-939-3
UN: 2922
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR