N,N-Dimethylethylendiamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N,N-Dimethylethylenediamine, asym. Dimethylethylendiamine, unsym-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, 2-Dimethylaminoethylamine, 2-Aminoethyldimethylamine
39030-100ML 39030-500ML
SIAL39030-100MLEA 131 EUR
SIAL39030-100ML SIAL39030-500ML
N,N-Dimethylethylendiamine, Sigma-Aldrich®
N,N-Dimethylethylendiamine
Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: ∼106 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.804 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00008175
CAS Number: 108-00-9
EINECS: 203-541-2