2-Diethylaminoethanol, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
2-(Diethylamino)-ethanol, N,N,-Diethylethanolamine, N,N-Diethylethanolamine
M-Clarity™ quality level = MQ100
471321-100ML
471321-18L
471321-2L
471321-500ML
SIAL471321-100MLEA
37
EUR
SIAL471321-100ML
SIAL471321-18L
SIAL471321-2L
SIAL471321-500ML
2-Diethylaminoethanol, Sigma-Aldrich®
2-Diethylaminoethanol
Formula:
C₆H₁₅NO MW: 117.19 g/mol Boiling Pt: 162 °C (1013 hPa) Melting Pt: –70 °C Density: 0.8921 g/cm³ (20 °C) Flash Pt: 52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002850 CAS Number: 100-37-8 EINECS: 202-845-2 UN: 2686 ADR: 8,II Merck Index: 13,03139 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...