2-(Dimethylamino)-ethanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Dimethylaminoethanol, N,N-Dimethylethanolamine, DMAE
M-Clarity™ quality level = MQ100
38990-1L 38990-250ML
SIAL38990-1LEA 50.7 EUR
SIAL38990-1L SIAL38990-250ML
2-(Dimethylamino)-ethanol, Sigma-Aldrich®
2-Dimethylaminoethanol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 132…135 °C (1013 hPa)
Melting Pt: –59 °C
Density: 0.8871 g/cm³ (20 °C)
Flash Pt: 39
Storage Temperature: Ambient
MDL Number: MFCD00002846
CAS Number: 108-01-0
EINECS: 203-542-8
UN: 2051
ADR: 8 II
Merck Index: 13,02863