2-(Dimethylamino)-ethanol, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
2-Dimethylaminoethanol, N,N-Dimethylethanolamine, DMAE
M-Clarity™ quality level = MQ200
471453-100ML
471453-2L
471453-500ML
SIAL471453-100MLEA
33.4
EUR
SIAL471453-100ML
SIAL471453-2L
SIAL471453-500ML
2-(Dimethylamino)-ethanol, Sigma-Aldrich®
2-Dimethylaminoethanol
Formula:
C₄H₁₁NO MW: 89.14 g/mol Boiling Pt: 132…135 °C (1013 hPa) Melting Pt: –59 °C Density: 0.8871 g/cm³ (20 °C) Flash Pt: 39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002846 CAS Number: 108-01-0 EINECS: 203-542-8 UN: 2051 ADR: 8,II Merck Index: 13,02863 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...