1,4-Diisocyanatobutane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Tetramethylene diisocyanate
M-Clarity™ quality level = MQ200
371130-1G 371130-5G
SIAL371130-1GEA 128 EUR
SIAL371130-1G SIAL371130-5G
1,4-Diisocyanatobutane, Sigma-Aldrich®
1,4-Diisocyanatobutane
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Boiling Pt: 108…110 °C (15 mmHg)
Melting Pt: 185…187 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00075559
CAS Number: 4538-37-8
UN: 3334
ADR: 9,