1,6-Diisocyanatohexane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Hexamethylene diisocyanate
M-Clarity™ quality level = MQ200
52650-1L 52650-250ML
SIAL52650-1LEA 192 EUR
SIAL52650-1L SIAL52650-250ML
1,6-Diisocyanatohexane, Sigma-Aldrich®
1,6-Diisocyanatohexane
Formula: C₈H₁₂N₂O₂
MW: 168.2 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: –67 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00002047
CAS Number: 822-06-0
EINECS: 212-485-8
UN: 2281
ADR: 6.1,II