1-Butyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Butyl isothiocyanate
M-Clarity™ quality level = MQ200
253790-25G 253790-5G
SIAL253790-25GEA 208 EUR
SIAL253790-25G SIAL253790-5G
1-Butyl isothiocyanate, Sigma-Aldrich®
1-Butyl isothiocyanate
Formula: C₅H₉NS
MW: 115.2 g/mol
Boiling Pt: 66 °C (1013 hPa)
Density: 0.9559 g/cm³ (11.2 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004824
CAS Number: 592-82-5
EINECS: 209-770-4
UN: 1693
ADR: 6.1,II