Ethyl isothiocyanate 96%
Supplier: Thermo Fisher Scientific
Danger
376860050 376860250 376861000
ACRO376860050EA 19.7 EUR
ACRO376860050 ACRO376860250 ACRO376861000
Ethyl isothiocyanate 96%
Ethyl isothiocyanate
Formula: C₃H₅NS
MW: 87.15 g/mol
Boiling Pt: 130…132 °C (760 mmHg)
Melting Pt: –6 °C
Density: 0.995 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00004820
CAS Number: 542-85-8
EINECS: 208-831-2
UN: 3286
ADR: 3 II
Merck Index: 13,03848
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5140 (20°C, 589 nm)
GC ≥95 %