Butylamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1-Butylamine, 1-Aminobutane, n-Butylamine
M-Clarity™ quality level = MQ200
471305-250ML
471305-25ML
471305-1L
SIAL471305-250MLEA
42.2
EUR
SIAL471305-250ML
SIAL471305-25ML
SIAL471305-1L
Butylamine, Sigma-Aldrich®
Butylamine
Formula:
H₃C(CH₂)₂CH₂NH₂ MW: 73.14 g/mol Boiling Pt: 77 °C (1013 hPa) Melting Pt: –50 °C Density: 0.741 g/cm³ (20 °C) Flash Pt: –8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011690 CAS Number: 109-73-9 EINECS: 203-699-2 UN: 1125 ADR: 3,II Merck Index: 12,01578 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...