Octadecylamine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIALO1408-500GEA 72.9 EUR
SIALO1408-500G SIALO1408-3KG
Octadecylamine, Sigma-Aldrich®
Octadecylamine
Formula: H₃C(CH₂)₁₆CH₂NH₂
MW: 269,51 g/mol
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 50…52 °C
Density: 0,8618 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008159
UN: 3077
ADR: 9,III