Pentylamine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Amylamine, 1-Aminopentane, n-Amylamine, n-Pentylamine, 1-Pentylamine
M-Clarity™ quality level = MQ300
W424201-100G
W424201-1KG
W424201-SAMPLE
SIALW424201-100GEA
326
EUR
SIALW424201-100G
SIALW424201-1KG
SIALW424201-SAMPLE
Pentylamine, Sigma-Aldrich®
Pentylamine
Formula:
H₃C(CH₂)₃CH₂NH₂ MW: 87.16 g/mol Boiling Pt: 104.4 °C (1013 hPa) Melting Pt: –50 °C Density: 0.7547 g/cm³ (20 °C) Flash Pt: 7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008236 CAS Number: 110-58-7 EINECS: 203-780-2 UN: 1106 ADR: 3,II Merck Index: 13,00602 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...