Propylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Propylamine, 1-Aminopropane
M-Clarity™ quality level = MQ100
109819-1L 109819-250ML
SIAL109819-1LEA 80.9 EUR
SIAL109819-1L SIAL109819-250ML
Propylamine, Sigma-Aldrich®
Propylamine
Formula: H₃CCH₂CH₂NH₂
MW: 59.11 g/mol
Boiling Pt: 48 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.7175 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00008205
CAS Number: 107-10-8
EINECS: 203-462-3
UN: 1277
ADR: 3,II
Merck Index: 12,08028