Isopropylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Aminopropane
M-Clarity™ quality level = MQ200
471291-250ML 471291-25ML 471291-18L 471291-1L 471291-2L
SIAL471291-250MLEA 32.5 EUR
SIAL471291-250ML SIAL471291-25ML SIAL471291-18L SIAL471291-1L SIAL471291-2L
Isopropylamine, Sigma-Aldrich®
Isopropylamine
Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 32.4 °C (1013 hPa)
Melting Pt: –101 °C
Density: 0.69 g/cm³ (20 °C)
Flash Pt: –27 °C
Storage Temperature: Ambient
MDL Number: MFCD00008082
CAS Number: 75-31-0
EINECS: 200-860-9
UN: 1221
ADR: 3,I
Merck Index: 13,05229