Ethanolamine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL15014-1LEA 65.1 EUR
SIAL15014-1L
Ethanolamine, Sigma-Aldrich®
Ethanolamine
Formula: NH₂CH₂CH₂OH
MW: 61,08 g/mol
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: 10,5 °C
Density: 1,0164 g/cm³ (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00008183
CAS Number: 141-43-5
UN: 2491
ADR: 8,III
Merck Index: 13,03762
https://si.vwr.com/stibo/web/std.lang.all/11/67/2301167.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR