Ethanolamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
398136-500ML
398136-2.5L
398136-25ML
SIAL398136-500MLEA
68
EUR
SIAL398136-500ML
SIAL398136-2.5L
SIAL398136-25ML
Ethanolamine, Sigma-Aldrich®
Ethanolamine
Formula:
NH₂CH₂CH₂OH Boiling Pt: 171 °C (1013 hPa) Melting Pt: 10.5 °C Density: 1.0164 g/cm³ (20 °C) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008183 CAS Number: 141-43-5 EINECS: 205-483-3 UN: 2491 ADR: 8,III Merck Index: 13,03762 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...