4-Aminobutan-1-ol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 4-Hydroxybutylamine, 4-Amino-1-butanol

M-Clarity™ quality level = MQ200

178330-1G 178330-5G
SIAL178330-1GEA 39.2 EUR
SIAL178330-1G SIAL178330-5G
4-Aminobutan-1-ol, Sigma-Aldrich®
4-Aminobutan-1-ol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 206
Melting Pt: 16…18 °C
Density: 0.967 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008230
CAS Number: 13325-10-5
EINECS: 236-364-4
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR